Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001266997.1 | NP_001253926.1 | - | 461 | 106 |
XM_028849128.1 | XP_028704961.1 | - | 478 | 106 | |
XM_028849129.1 | XP_028704962.1 | - | 304 | 106 | |
GenBank mRNA | JU323472.1 | - | - | 461 | 106 |