Assay Design



Gene Transcripts

Gene Symbol
SLC7A11
Entrez Gene ID
696516
Gene Name
solute carrier family 7 member 11
Gene Aliases
-
Chromosome Location
Chr.5: 136037076 - 136110133 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266291.1 NP_001253220.1 - 1277 71
GenBank mRNA JU321205.1 - - 1277 71

View Full Product Details