Assay Design



Gene Transcripts

Gene Symbol
N4BP3
Entrez Gene ID
701704
Gene Name
NEDD4 binding protein 3
Gene Aliases
-
Chromosome Location
Chr.6: 175438894 - 175451194 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001093839.4 XP_001093839.1 - 675 87
GenBank mRNA JV610127.1 - - 432 87

View Full Product Details