Assay Design



Gene Transcripts

Gene Symbol
FBXO28
Entrez Gene ID
702246
Gene Name
F-box protein 28
Gene Aliases
-
Chromosome Location
Chr.1: 81572430 - 81622804 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001097187.4 XP_001097187.1 - 762 145
GenBank mRNA JU326704.1 - - 705 145
JV710422.1 - - 708 145

View Full Product Details