Assay Design



Gene Transcripts

Gene Symbol
NUDCD3
Entrez Gene ID
699353
Gene Name
NudC domain containing 3
Gene Aliases
EGK_13730
Chromosome Location
Chr.3: 70728249 - 70834677 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266593.1 NP_001253522.1 - 961 67
GenBank mRNA JU333671.1 - - 989 67
JU474924.1 - - 988 67
JV046582.1 - - 961 67
JV710772.1 - - 990 67

View Full Product Details