Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_015116253.2 | XP_014971739.1 | - | 1951 | 114 |
GenBank mRNA | JU333872.1 | - | - | 305 | 114 |
JU333873.1 | - | - | 305 | 114 | |
JU472033.1 | - | - | 326 | 114 | |
JU472034.1 | - | - | 326 | 114 | |
JV046684.1 | - | - | 456 | 114 | |
JV046685.1 | - | - | 456 | 114 |