Assay Design



Gene Transcripts

Gene Symbol
CCDC69
Entrez Gene ID
713304
Gene Name
coiled-coil domain containing 69
Gene Aliases
-
Chromosome Location
Chr.6: 148631782 - 148674690 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260862.1 NP_001247791.1 - 348 84
XM_015141196.2 XP_014996682.1 - 456 84
GenBank mRNA JU472300.1 - - 348 84
JU475109.1 - - 348 84
JV057555.1 - - 202 84

View Full Product Details