Assay Design



Gene Transcripts

Gene Symbol
HIGD1B
Entrez Gene ID
715566
Gene Name
HIG1 hypoxia inducible domain family member 1B
Gene Aliases
EGK_08815
Chromosome Location
Chr.16: 56116010 - 56119170 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015119890.2 XP_014975376.1 - 391 91
XM_015119891.2 XP_014975377.1 - 349 91
XM_028836607.1 XP_028692440.1 - 449 91
GenBank mRNA JU323424.1 - - 337 91

View Full Product Details