Assay Design



Gene Transcripts

Gene Symbol
LAX1
Entrez Gene ID
702011
Gene Name
lymphocyte transmembrane adaptor 1
Gene Aliases
-
Chromosome Location
Chr.1: 61272534 - 61283965 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001101278.4 XP_001101278.1 - 752 120
GenBank mRNA JU471241.1 - - 723 120
JU476146.1 - - 723 120

View Full Product Details