Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_015149557.2 | XP_015005043.2 | - | 868 | 144 |
XM_015149559.2 | XP_015005045.2 | - | 938 | 144 | |
XM_028828148.1 | XP_028683981.1 | - | 1003 | 144 | |
XM_028828149.1 | XP_028683982.1 | - | 894 | 144 | |
XM_028828150.1 | XP_028683983.1 | - | 1403 | 144 | |
XM_028828151.1 | XP_028683984.1 | - | 1385 | 144 | |
XM_028828152.1 | XP_028683985.1 | - | 1460 | 144 | |
XM_028828153.1 | XP_028683986.1 | - | 1460 | 144 | |
XM_028828154.1 | XP_028683987.1 | - | 1460 | 144 | |
XM_028828156.1 | XP_028683989.1 | - | 1460 | 144 |