Assay Design



Gene Transcripts

Gene Symbol
TRIM62
Entrez Gene ID
709927
Gene Name
tripartite motif containing 62
Gene Aliases
-
Chromosome Location
Chr.1: 191466769 - 191501834 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257752.1 NP_001244681.1 - 1499 110
GenBank mRNA JU335273.1 - - 1499 110

View Full Product Details