Assay Design



Gene Transcripts

Gene Symbol
DOK5
Entrez Gene ID
698689
Gene Name
docking protein 5
Gene Aliases
-
Chromosome Location
Chr.10: 89681758 - 89854483 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260578.1 NP_001247507.1 - 328 117
XM_015148745.2 XP_015004231.1 - 306 117
XM_028827209.1 XP_028683042.1 - 646 117
GenBank mRNA JU335440.1 - - 328 117
JV550831.1 - - 376 117
JV587202.1 - - 364 117

View Full Product Details