Assay Design



Gene Transcripts

Gene Symbol
CASS4
Entrez Gene ID
697924
Gene Name
Cas scaffold protein family member 4
Gene Aliases
C20orf32
Chromosome Location
Chr.10: 91551662 - 91605369 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015148735.2 XP_015004221.2 - 992 87
XM_028828521.1 XP_028684354.1 - 342 87

View Full Product Details