Assay Design



Gene Transcripts

Gene Symbol
CLTRN
Entrez Gene ID
712928
Gene Name
collectrin, amino acid transport regulator
Gene Aliases
EGK_20289, TMEM27
Chromosome Location
Chr.X: 15325117 - 15363203 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194195.2 NP_001181124.1 - 452 126

View Full Product Details