Assay Design



Gene Transcripts

Gene Symbol
SYT4
Entrez Gene ID
694191
Gene Name
synaptotagmin 4
Gene Aliases
EGK_09650
Chromosome Location
Chr.18: 39201210 - 39211054 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265986.1 NP_001252915.1 - 1095 129
GenBank mRNA JU321413.1 - - 1095 129

View Full Product Details