Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001194649.1 | NP_001181578.1 | - | 198 | 60 |
GenBank mRNA | JU320864.1 | - | - | 183 | 60 |
JV529545.1 | - | - | 198 | 60 | |
JV567959.1 | - | - | 207 | 60 | |
JV668791.1 | - | - | 200 | 60 |