Assay Design



Gene Transcripts

Gene Symbol
GDAP1L1
Entrez Gene ID
693773
Gene Name
ganglioside induced differentiation associated protein 1 like 1
Gene Aliases
EGK_02390
Chromosome Location
Chr.10: 79476171 - 79509012 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257531.1 NP_001244460.1 - 791 132
GenBank mRNA JU336331.1 - - 791 132
JV667993.1 - - 830 132

View Full Product Details