Assay Design



Gene Transcripts

Gene Symbol
METTL22
Entrez Gene ID
708739
Gene Name
methyltransferase like 22
Gene Aliases
C16orf68
Chromosome Location
Chr.20: 8673539 - 8699653 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194202.2 NP_001181131.2 - 706 98
XM_015125570.2 XP_014981056.2 - 1016 98
XM_015125571.2 XP_014981057.2 - 1029 98
XM_015125573.2 XP_014981059.2 - 1016 98
XM_028840519.1 XP_028696352.1 - 1016 98
GenBank mRNA JU323753.1 - - 942 98
JU472906.1 - - 830 98

View Full Product Details