Assay Design



Gene Transcripts

Gene Symbol
FN3KRP
Entrez Gene ID
716431
Gene Name
fructosamine 3 kinase related protein
Gene Aliases
-
Chromosome Location
Chr.16: 79066861 - 79079754 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194815.3 NP_001181744.2 - 626 114
GenBank mRNA JU336402.1 - - 643 114
JU477220.1 - - 611 114
JV048260.1 - - 626 114

View Full Product Details