Assay Design



Gene Transcripts

Gene Symbol
B9D2
Entrez Gene ID
704873
Gene Name
B9 domain containing 2
Gene Aliases
EGK_10650
Chromosome Location
Chr.19: 40599375 - 40610003 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015124021.2 XP_014979507.2 - 1115 78
XM_015124024.2 XP_014979510.1 - 324 78
GenBank mRNA JU336516.1 - - 295 78

View Full Product Details