Assay Design



Gene Transcripts

Gene Symbol
YIPF5
Entrez Gene ID
706656
Gene Name
Yip1 domain family member 5
Gene Aliases
-
Chromosome Location
Chr.6: 141554762 - 141569048 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260957.1 NP_001247886.1 - 169 88
GenBank mRNA JU332312.1 - - 109 88
JU332313.1 - - 109 88
JU332314.1 - - 109 88
JU332315.1 - - 109 88
JU332316.1 - - 109 88
JU472191.1 - - 130 88
JV045670.1 - - 169 88

Gene Symbol
LOC114675375
Entrez Gene ID
114675375
Gene Name
protein YIPF5-like
Gene Aliases
-
Chromosome Location
Chr.6: 4064 - 10476 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028842634.1 XP_028698467.1 - 142 88

View Full Product Details