Assay Design



Gene Transcripts

Gene Symbol
YIPF5
Entrez Gene ID
706656
Gene Name
Yip1 domain family member 5
Gene Aliases
-
Chromosome Location
Chr.6: 141554762 - 141569048 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260957.1 NP_001247886.1 - 793 125
XM_015141101.2 XP_014996587.1 - 1079 125
GenBank mRNA JU332312.1 - - 733 125
JU332313.1 - - 733 125
JU332314.1 - - 733 125
JU332315.1 - - 733 125
JU332316.1 - - 733 125
JU472191.1 - - 754 125
JV045670.1 - - 793 125

View Full Product Details