Assay Design



Gene Transcripts

Gene Symbol
RILP
Entrez Gene ID
701798
Gene Name
Rab interacting lysosomal protein
Gene Aliases
-
Chromosome Location
Chr.16: 1507411 - 1511260 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001086142.4 XP_001086142.1 - 505 115
XM_028835854.1 XP_028691687.1 - 505 115

View Full Product Details