Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_015118605.2 | XP_014974091.1 | - | 927 | 98 |
XM_015118606.2 | XP_014974092.1 | - | 742 | 98 | |
XM_015118607.2 | XP_014974093.1 | - | 1323 | 98 | |
XM_015118608.2 | XP_014974094.1 | - | 1189 | 98 | |
XM_015118609.2 | XP_014974095.1 | - | 927 | 98 | |
XM_015118610.2 | XP_014974096.1 | - | 742 | 98 | |
GenBank mRNA | JU336277.1 | - | - | 648 | 98 |
JU477119.1 | - | - | 673 | 98 |