Assay Design



Gene Transcripts

Gene Symbol
IZUMO2
Entrez Gene ID
719187
Gene Name
IZUMO family member 2
Gene Aliases
C19orf41
Chromosome Location
Chr.19: 50054665 - 50066945 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194696.1 NP_001181625.1 - 401 124

View Full Product Details