Assay Design



Gene Transcripts

Gene Symbol
IQCF1
Entrez Gene ID
699719
Gene Name
IQ motif containing F1
Gene Aliases
EGK_11582
Chromosome Location
Chr.2: 108023088 - 108029740 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001095004.4 XP_001095004.2 - 282 130

View Full Product Details