Assay Design



Gene Transcripts

Gene Symbol
MS4A15
Entrez Gene ID
697157
Gene Name
membrane spanning 4-domains A15
Gene Aliases
-
Chromosome Location
Chr.14: 13435842 - 13455105 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001085764.4 XP_001085764.3 - 617 117

View Full Product Details