Assay Design



Gene Transcripts

Gene Symbol
CYP4F22
Entrez Gene ID
718303
Gene Name
cytochrome P450, family 4, subfamily F, polypeptide 22
Gene Aliases
-
Chromosome Location
Chr.19: 15106415 - 15149547 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194539.2 NP_001181468.2 - 1470 78

View Full Product Details