Assay Design



Gene Transcripts

Gene Symbol
TRIM65
Entrez Gene ID
708355
Gene Name
tripartite motif containing 65
Gene Aliases
-
Chromosome Location
Chr.16: 72259822 - 72267787 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028836809.1 XP_028692642.1 - 1072 104
GenBank mRNA JU482164.1 - - 705 104
JV709053.1 - - 65 104

View Full Product Details