Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_001113056.4 | XP_001113056.3 | - | 2395 | 119 |
XM_028845591.1 | XP_028701424.1 | - | 2807 | 119 | |
GenBank mRNA | JU324067.1 | - | - | 2107 | 119 |
JV645403.1 | - | - | 2108 | 119 |