Assay Design



Gene Transcripts

Gene Symbol
BPIFC
Entrez Gene ID
717287
Gene Name
BPI fold containing family C
Gene Aliases
BPIL2, EGK_02973
Chromosome Location
Chr.10: 19033699 - 19094573 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001112428.3 XP_001112428.1 - 1525 154
XM_028827763.1 XP_028683596.1 - 1792 154
GenBank mRNA JV711263.1 - - 172 154

View Full Product Details