Assay Design



Gene Transcripts

Gene Symbol
SLC46A3
Entrez Gene ID
712056
Gene Name
solute carrier family 46 member 3
Gene Aliases
-
Chromosome Location
Chr.17: 7904642 - 7922616 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015120809.2 XP_014976295.2 - 1835 85
XM_028837533.1 XP_028693366.1 - 2295 85

View Full Product Details