Assay Design



Gene Transcripts

Gene Symbol
VMO1
Entrez Gene ID
709950
Gene Name
vitelline membrane outer layer 1 homolog
Gene Aliases
-
Chromosome Location
Chr.16: 4616451 - 4617516 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194053.2 NP_001180982.2 - 316 138
GenBank mRNA JU342097.1 - - 384 138
JU478354.1 - - 345 138

View Full Product Details