Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_001097091.4 | XP_001097091.1 | - | 453 | 130 |
XM_001097205.4 | XP_001097205.1 | - | 409 | 130 | |
XM_028834614.1 | XP_028690447.1 | - | 454 | 130 | |
XM_028834615.1 | XP_028690448.1 | - | 643 | 130 |