Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_015140399.2 | XP_014995885.1 | - | 266 | 184 |
XM_015140401.2 | XP_014995887.1 | - | 267 | 184 | |
GenBank mRNA | JU474233.1 | - | - | 142 | 184 |
JV046126.1 | - | - | 141 | 184 |