Assay Design



Gene Transcripts

Gene Symbol
AADACL2
Entrez Gene ID
709222
Gene Name
arylacetamide deacetylase like 2
Gene Aliases
EGK_11992
Chromosome Location
Chr.2: 53959145 - 53986547 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001106757.2 XP_001106757.2 - 256 111

View Full Product Details