Assay Design



Gene Transcripts

Gene Symbol
CRP
Entrez Gene ID
719454
Gene Name
C-reactive protein
Gene Aliases
EGK_01483
Chromosome Location
Chr.1: 91224567 - 91226972 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001117250.4 XP_001117250.2 - 321 74

View Full Product Details