Assay Design



Gene Transcripts

Gene Symbol
PTGDR
Entrez Gene ID
707918
Gene Name
prostaglandin D2 receptor
Gene Aliases
-
Chromosome Location
Chr.7: 114391661 - 114404290 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001103541.4 XP_001103541.1 - 952 111
GenBank mRNA JU490122.1 - - 774 111

View Full Product Details