Assay Design



Gene Transcripts

Gene Symbol
OR52W1
Entrez Gene ID
713141
Gene Name
olfactory receptor, family 52, subfamily W, member 1
Gene Aliases
EGK_06549
Chromosome Location
Chr.14: 59744667 - 59745773 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194213.1 NP_001181142.1 - 730 72

View Full Product Details