Assay Design



Gene Transcripts

Gene Symbol
BCL2L15
Entrez Gene ID
707537
Gene Name
BCL2 like 15
Gene Aliases
-
Chromosome Location
Chr.1: 110326808 - 110334373 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001110407.4 XP_001110407.2 - 808 106

View Full Product Details