Assay Design



Gene Transcripts

Gene Symbol
BMP2
Entrez Gene ID
718330
Gene Name
bone morphogenetic protein 2
Gene Aliases
EGK_02657
Chromosome Location
Chr.10: 59734131 - 59746413 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001115987.3 XP_001115987.1 - 1201 87
GenBank mRNA JV659136.1 - - 156 87

View Full Product Details