Assay Design



Gene Transcripts

Gene Symbol
UCN
Entrez Gene ID
699942
Gene Name
urocortin
Gene Aliases
-
Chromosome Location
Chr.13: 81643721 - 81644580 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265661.1 NP_001252590.1 - 96 66
GenBank mRNA JV600234.1 - - 96 66

View Full Product Details