Assay Design



Gene Transcripts

Gene Symbol
NPBWR2
Entrez Gene ID
719200
Gene Name
neuropeptides B and W receptor 2
Gene Aliases
EGK_02162
Chromosome Location
Chr.10: 99332396 - 99335406 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001113051.4 XP_001113051.2 - 475 109

View Full Product Details