Assay Design



Gene Transcripts

Gene Symbol
NEUROG1
Entrez Gene ID
711992
Gene Name
neurogenin 1
Gene Aliases
-
Chromosome Location
Chr.6: 132995715 - 132997436 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001110622.4 XP_001110622.1 - 1531 123

View Full Product Details