Assay Design



Gene Transcripts

Gene Symbol
HOXB13
Entrez Gene ID
697572
Gene Name
homeobox B13
Gene Aliases
-
Chromosome Location
Chr.16: 48072081 - 48074569 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001089620.4 XP_001089620.2 - 883 108

View Full Product Details