Assay Design



Gene Transcripts

Gene Symbol
CHSY1
Entrez Gene ID
694481
Gene Name
chondroitin sulfate synthase 1
Gene Aliases
-
Chromosome Location
Chr.7: 80463571 - 80538154 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001084554.4 XP_001084554.4 - 830 143
GenBank mRNA JV636035.1 - - 850 143
JV709788.1 - - 567 143

View Full Product Details