Assay Design



Gene Transcripts

Gene Symbol
HIST1H2AD
Entrez Gene ID
697235
Gene Name
histone cluster 1, H2ad
Gene Aliases
EGK_14555
Chromosome Location
Chr.4: 143791245 - 143791637 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193793.1 NP_001180722.1 - 244 109

View Full Product Details