Assay Design



Gene Transcripts

Gene Symbol
DUSP3
Entrez Gene ID
713686
Gene Name
dual specificity phosphatase 3
Gene Aliases
-
Chromosome Location
Chr.16: 55009755 - 55020603 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266548.1 NP_001253477.1 - 164 146
GenBank mRNA JU333262.1 - - 191 146
JV046357.1 - - 164 146

View Full Product Details