Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257627.1 | NP_001244556.1 | - | 1002 | 137 |
XM_015133733.2 | XP_014989219.2 | - | 1079 | 137 | |
GenBank mRNA | JU335814.1 | - | - | 1002 | 137 |
JV047864.1 | - | - | 1034 | 137 | |
JV047866.1 | - | - | 1025 | 137 |