Assay Design



Gene Transcripts

Gene Symbol
CCR3
Entrez Gene ID
713368
Gene Name
C-C motif chemokine receptor 3
Gene Aliases
CMKBR3
Chromosome Location
Chr.2: 102268893 - 102269960 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047140.1 NP_001040605.1 - 757 120

View Full Product Details